Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylbenzo[b]thiophene, 98%

Catalog No. AAA1577206
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 1455-18-1 | C9H8S | 148.22 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Notes
participates in base-catalyzed Knoevenagel condensation reaction
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1455-18-1
Molecular Formula C9H8S
Molecular Weight (g/mol) 148.22
MDL Number MFCD00040243
InChI Key SEBRPHZZSLCDRQ-UHFFFAOYSA-N
Synonym 3-methylbenzo b thiophene, 3-methylbenzothiophene, 3-methylthianaphthene, benzo b thiophene, 3-methyl, 3-methylthioindene, benzothiophene, 3-methyl, 3-methyl-1-thiaindene, 3-methylbenzylthiophene, 3-methyl-benzo b thiophene
PubChem CID 73817
IUPAC Name 3-methyl-1-benzothiophene
SMILES CC1=CSC2=CC=CC=C12

Specifications

Melting Point -40°C
Density 1.13
Boiling Point 72°C to 74°C (2 mmHg)
Flash Point 104°C (219°F)
Refractive Index 1.624
Quantity 5 g
Beilstein 111282
Formula Weight 148.23
Percent Purity 98%
Chemical Name or Material 3-Methylbenzo[b]thiophene
EINECSNumber 215-934-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.