Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Methylbenzyl bromide, 97%

Click to view available options
Quantity:
5g
25g
620-13-3
C8H9Br
185.064
MFCD00000177
FWLWTILKTABGKQ-UHFFFAOYSA-N
3-methylbenzyl bromide,1-bromomethyl-3-methylbenzene,alpha-bromo-m-xylene,m-methylbenzyl bromide,m-xylyl bromide,benzene, 1-bromomethyl-3-methyl,3-bromomethyl toluene,m-xylene, .alpha.-bromo,3-methylbenzylbromide,m-xylene, alpha-bromo
12099
1-(bromomethyl)-3-methylbenzene
CC1=CC(=CC=C1)CBr
620-13-3
C8H9Br
185.064
MFCD00000177
FWLWTILKTABGKQ-UHFFFAOYSA-N
3-methylbenzyl bromide,1-bromomethyl-3-methylbenzene,alpha-bromo-m-xylene,m-methylbenzyl bromide,m-xylyl bromide,benzene, 1-bromomethyl-3-methyl,3-bromomethyl toluene,m-xylene, .alpha.-bromo,3-methylbenzylbromide,m-xylene, alpha-bromo
12099
1-(bromomethyl)-3-methylbenzene
CC1=CC(=CC=C1)CBr
Refractive Index 1.569
Density 1.37
Quantity 5g
Boiling Point 184°C to 185°C (340mmHg)
Flash Point 82°C (179°F)
UN Number UN1701
Beilstein 2076425
Merck Index 14,10090
Formula Weight 185.07
Percent Purity 97%
Chemical Name or Material 3-Methylbenzyl bromide
Show More Show Less

GHS H Statement
H330-H314-H318-H227
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H227-H302+H312+H332-H314

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: XYLYL BROMIDE, LIQUID

EINECSNumber : 210-625-2

RTECSNumber : CY9039200

TSCA : Yes

Recommended Storage : Keep cold