Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylcyclohexanone 97%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass bottle
591-24-2
C7H12O
112.17
MFCD00001639
UJBOOUHRTQVGRU-UHFFFAOYSA-N
3-methylcyclohexanone,cyclohexanone, 3-methyl,methyl-3 cyclohexanone-1,3-methyl cyclohexanone,methyl-3 cyclohexanone-1 french,m-methylcyclohexanone,r-3-methylcyclohexanone,+-3r-3-methylcyclohexanone,cyclohexanone, 3-methyl-, r,cyclohexanone, 3-methyl-, s
11567
3-methylcyclohexan-1-one
CC1CCCC(=O)C1
591-24-2
C7H12O
112.17
MFCD00001639
UJBOOUHRTQVGRU-UHFFFAOYSA-N
3-methylcyclohexanone,cyclohexanone, 3-methyl,methyl-3 cyclohexanone-1,3-methyl cyclohexanone,methyl-3 cyclohexanone-1 french,m-methylcyclohexanone,r-3-methylcyclohexanone,+-3r-3-methylcyclohexanone,cyclohexanone, 3-methyl-, r,cyclohexanone, 3-methyl-, s
11567
3-methylcyclohexan-1-one
CC1CCCC(=O)C1
Quantity 5g
Boiling Point 169.0°C to 170.0°C
Density 0.9150g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4440 to 1.4460
Melting Point -73.0°C
Color Colorless to Yellow
Specific Gravity 0.915
Flash Point 48°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH3C6H9(=O)
Beilstein 07, 15
Solubility Information Solubility in water: insoluble
Formula Weight 112.17
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 3-Methylcyclohexanone, 97%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 209-710-7

RTECSNumber : GW1750100

TSCA : TSCA