Learn More
Invitrogen™ 3-Methylcytosine Polyclonal Antibody
Description
3-Methylcytosine Polyclonal Antibody for ARRAY
Specifications
Specifications
| Antigen | 3-Methylcytosine |
| Applications | Peptide Array |
| Classification | Polyclonal |
| Concentration | 1.8 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% ProClin 300, 0.05% sodium azide; pH 7.4 |
| Gene Alias | 3mc; 3-mC; 3-meC; 3-Methyl-cytosine; C5H7NO3 |
| Host Species | Rabbit |
| Immunogen | Human BSA-conjugated 3-methylcytosine (3-mC). |
| Purification Method | Protein G |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.