Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylglutaric anhydride, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
4166-53-4
C6H8O3
128.13
MFCD00006683
MGICRVTUCPFQQZ-UHFFFAOYSA-N
3-methylglutaric anhydride,4-methyldihydro-2h-pyran-2,6 3h-dione,3-methylglutaricanhydride,.beta.-methylglutaric anhydride,acmc-1ago6,2h-pyran-2,6 3h-dione, dihydro-4-methyl,3-methyl glutaric anhydride,beta-methylglutaric anhydride,4-methyltetrahydropyran-2,6-dione
77828
4-methyloxane-2,6-dione
CC1CC(=O)OC(=O)C1
4166-53-4
C6H8O3
128.13
MFCD00006683
MGICRVTUCPFQQZ-UHFFFAOYSA-N
3-methylglutaric anhydride,4-methyldihydro-2h-pyran-2,6 3h-dione,3-methylglutaricanhydride,.beta.-methylglutaric anhydride,acmc-1ago6,2h-pyran-2,6 3h-dione, dihydro-4-methyl,3-methyl glutaric anhydride,beta-methylglutaric anhydride,4-methyltetrahydropyran-2,6-dione
77828
4-methyloxane-2,6-dione
CC1CC(=O)OC(=O)C1
Boiling Point 180°C to 182°C (25.0mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5g
Melting Point 40°C to 45°C
Color White
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 17, 415
Solubility Information Solubility in water: may decompose
Formula Weight 128.13
Physical Form Crystalline Low Melting Solid
Percent Purity 97%
Chemical Name or Material 3-Methylglutaric anhydride, 97%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 224-020-6

TSCA : TSCA