Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylpyrazole 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
1453-58-3
C4H6N2
82.1
MFCD00005240
XKVUYEYANWFIJX-UHFFFAOYSA-N
3-methylpyrazole,3-methyl-1h-pyrazole,1h-pyrazole, 3-methyl,5-methylpyrazole,3 5-methylpyrazole,pyrazole, 5-methyl,1h-pyrazole,5-methyl,1h-pyrazole, 5-methyl,pyrazole, 3-methyl,unii-w7ku1rro6u
15073
5-methyl-1H-pyrazole
CC1=CC=NN1
1453-58-3
C4H6N2
82.1
MFCD00005240
XKVUYEYANWFIJX-UHFFFAOYSA-N
3-methylpyrazole,3-methyl-1h-pyrazole,1h-pyrazole, 3-methyl,5-methylpyrazole,3 5-methylpyrazole,pyrazole, 5-methyl,1h-pyrazole,5-methyl,1h-pyrazole, 5-methyl,pyrazole, 3-methyl,unii-w7ku1rro6u
15073
5-methyl-1H-pyrazole
CC1=CC=NN1
Boiling Point 204°C
Quantity 25g
Flash Point 93°C
Specific Gravity 1.02
Color Yellow
Density 1.0200g/mL
Refractive Index 1.496
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Beilstein 23, 50
Solubility Information Solubility in water: miscible
Formula Weight 82.1
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material 3-Methylpyrazole, 99%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 215-925-7

RTECSNumber : UQ7340000

TSCA : TSCA