Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylpyridine-2-carboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. M11411G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M11411G 1 g
M1141100MG 100 mg
M11415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M11411G Supplier TCI America Supplier No. M11411G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4021-07-2
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00191553
InChI Key LMHIBYREWJHKNZ-UHFFFAOYSA-N
Synonym 3-methylpicolinic acid, 3-methyl-2-pyridinecarboxylic acid, 3-methyl-2-picolinic acid, 2-pyridinecarboxylic acid, 3-methyl, 3-methyl-a-picolinic acid, 3-methyl-alpha-picolinic acid, 3-methylpyridine-2-carboxylicacid, methylpyridincarbonsaure, pubchem14396, pubchem17659
PubChem CID 268757
IUPAC Name 3-methylpyridine-2-carboxylic acid
SMILES CC1=C(N=CC=C1)C(=O)O

Specifications

Melting Point 119°C
Color White
Quantity 1 g
Formula Weight 137.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methylpyridine-2-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.