Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylsalicylic acid, 99%

Catalog No. p-3758471
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle

CAS: 83-40-9 | C8H8O3 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83-40-9
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00002448
InChI Key WHSXTWFYRGOBGO-UHFFFAOYSA-N
Synonym 3-methylsalicylic acid, hydroxytoluic acid, o-cresotic acid, o-cresotinic acid, cresotic acid, o-homosalicylic acid, 2-hydroxy-m-toluic acid, 2,3-cresotinic acid, cresotinic acid, benzoic acid, 2-hydroxy-3-methyl
PubChem CID 6738
ChEBI CHEBI:20141
IUPAC Name 2-hydroxy-3-methylbenzoic acid
SMILES CC1=CC=CC(C(O)=O)=C1O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 164.0°C to 167.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula CH3C6H3(OH)CO2H
Quantity 5 g
Beilstein 10, 220
Merck Index 15, 2570
Solubility Information Solubility in water: 1.5g/L (20°C). Other solubilities: soluble in alkali hydroxides, soluble in chloroform, alcohol and ether
Formula Weight 152.15
Percent Purity 99%
Physical Form Fine Powder
Chemical Name or Material 3-Methylsalicylic acid
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of s
EINECSNumber 201-473-8

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.