Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Methylthio)propylamine, 97%, Thermo Scientific™

Catalog No. AC414930010
Click to view available options
:
1g; Glass bottle

Specifications

CAS 4104-45-4
CAS Min % 96.0
CAS Max % 100.0
Density 0.9600g/mL
Boiling Point 169.0°C
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C4H11NS
Refractive Index 1.4920 to 1.4940
Linear Formula CH3SCH2CH2CH2NH2
Quantity 1g
MDL Number MFCD00041898
Beilstein 04, IV, 1655
Specific Gravity 0.96
Synonym 3-methylthiopropylamine, 3-methylthio propylamine, 1-propanamine, 3-methylthio, 3-methylthiopropanamine, methylmercaptopropylamine, unii-1iap0cgd5s, 3-methylmercaptopropylamine, 3-methylthio-1-propanamine, 3-methylsulfanyl propan-1-amine, 1iap0cgd5s
InChI Key KKYSBGWCYXYOHA-UHFFFAOYSA-N
SMILES CSCCCN
IUPAC Name 3-methylsulfanylpropan-1-amine
Molecular Weight (g/mol) 105.2
PubChem CID 77743
ChEBI CHEBI:16696
Formula Weight 105.2
Percent Purity 97%
Chemical Name or Material 3-(Methylthio)propylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 223-875-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.