Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-methylthiophene-2-carbonyl Chloride, 98%, ACROS Organics™

Click to view available options
Quantity:
2g
10g
Packaging:
Glass bottle
61341-26-2
C6H5ClOS
160.62
MFCD00130091
AJSVPEVDFBYRCH-UHFFFAOYSA-N
3-methyl-2-thiophenecarbonyl chloride,3-methylthiophene-2-carbonylchloride,3-methyl-2-thenoyl chloride,2-3-methyl thiophene carboxylicacid chloride,pubchem5561,acmc-209mrl,3-methylthenoyl chloride,3-methyl-2-thenoylchloride,3-methyl-2-thienoyl chloride,3-methylthiophenecarbonylchloride
2737605
3-methylthiophene-2-carbonyl chloride
CC1=C(SC=C1)C(=O)Cl
61341-26-2
C6H5ClOS
160.62
MFCD00130091
AJSVPEVDFBYRCH-UHFFFAOYSA-N
3-methyl-2-thiophenecarbonyl chloride,3-methylthiophene-2-carbonylchloride,3-methyl-2-thenoyl chloride,2-3-methyl thiophene carboxylicacid chloride,pubchem5561,acmc-209mrl,3-methylthenoyl chloride,3-methyl-2-thenoylchloride,3-methyl-2-thienoyl chloride,3-methylthiophenecarbonylchloride
2737605
3-methylthiophene-2-carbonyl chloride
CC1=C(SC=C1)C(=O)Cl
Flash Point >110°C
Boiling Point 216°C to 220°C
Infrared Spectrum Authentic
Refractive Index 1.5830 to 1.585
Color Colorless to Yellow
Quantity 10g
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 160.62
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 3-Methylthiophene-2-carbonyl chloride
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger