Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylthiophene, 99+%, ACROS Organics™

Click to view available options
Quantity:
50mL
250mL
Packaging:
Glass bottle
616-44-4
C5H6S
98.17
MFCD00005470
QENGPZGAWFQWCZ-UHFFFAOYSA-N
thiophene, 3-methyl,3-thiotolene,beta-methylthiophene,poly,p3mt,3-methyl thiophene,3-methyl-thiophene,methyl-3-thiophene,ccris 2937,thiophene, 3-methyl-, homopolymer
12024
3-methylthiophene
CC1=CSC=C1
616-44-4
C5H6S
98.17
MFCD00005470
QENGPZGAWFQWCZ-UHFFFAOYSA-N
thiophene, 3-methyl,3-thiotolene,beta-methylthiophene,poly,p3mt,3-methyl thiophene,3-methyl-thiophene,methyl-3-thiophene,ccris 2937,thiophene, 3-methyl-, homopolymer
12024
3-methylthiophene
CC1=CSC=C1
Boiling Point 115.0°C to 117.0°C
Infrared Spectrum Authentic
Melting Point -69.0°C
Refractive Index 1.5180 to 1.5200
Packaging Glass bottle
Flash Point 11°C
Specific Gravity 1.016
Quantity 250mL
Density 1.0160g/mL
Color Colorless to Yellow
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Beilstein 17, 38
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,ether,aceton and benzene
Formula Weight 98.17
Physical Form Liquid
Percent Purity ≥99%
Chemical Name or Material 3-Methylthiophene
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Causes skin irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger