Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylxanthine, 97%, Thermo Scientific™

Catalog No. AC206930010
Click to view available options
Quantity:
100 mg
250 mg
1 g

Chemical Identifiers

CAS 1076-22-8
Molecular Formula C6H6N4O2
Molecular Weight (g/mol) 166.14
InChI Key GMSNIKWWOQHZGF-UHFFFAOYSA-N
Synonym 3-methylxanthine, 3-methyl-1h-purine-2,6 3h,7h-dione, 2,6-dihydroxy-3-methylpurine, xanthine, 3-methyl, 3 mx, 3-methyl xanthine, 1h-purine-2,6-dione, 3,7-dihydro-3-methyl, unii-ws6x982oec, 3-methyl-2,3,6,7-tetrahydro-1h-purine-2,6-dione, ccris 5817
PubChem CID 70639
ChEBI CHEBI:62207
IUPAC Name 3-methyl-7H-purine-2,6-dione
SMILES CN1C2=C(C(=O)NC1=O)NC=N2

Specifications

Melting Point >300.0°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Beilstein 26,453
Solubility Information Solubility in water: insoluble
Formula Weight 166.14
Percent Purity 97%
Chemical Name or Material 3-Methylxanthine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
EINECSNumber 214-058-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.