Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Morpholinopropanesulfonic Acid 98.0+%, TCI America™
SDP

Catalog No. M070725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M070725G 25 g
M0707250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M070725G Supplier TCI America Supplier No. M070725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1132-61-2
Molecular Formula C7H15NO4S
Molecular Weight (g/mol) 209.26
MDL Number MFCD00006183
InChI Key DVLFYONBTKHTER-UHFFFAOYSA-N
Synonym mops, 4-morpholinepropanesulfonic acid, 3-morpholinopropanesulfonic acid, 3-n-morpholino propanesulfonic acid, 3-morpholinopropane-1-sulfonic acid, morpholinopropane sulfonic acid, morpholinopropanesulfonic acid, 4-morpholinepropane sulfonic acid, 3-n-morpholinopropanesulfonic acid, unii-273bp63nv3
PubChem CID 70807
ChEBI CHEBI:44115
IUPAC Name 4-(3-sulfonatopropyl)morpholin-4-ium
SMILES [O-]S(=O)(=O)CCC[NH+]1CCOCC1

Specifications

Color White
Quantity 25 g
Formula Weight 209.26
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Morpholinopropanesulfonic Acid [Good′s buffer component for biological research]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.