Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(N-BOC)-(2-aminoethyl)azetidine, 96%, Thermo Scientific™

Catalog No. AC451242500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC451242500 250 mg
AC451240010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC451242500 Supplier Thermo Scientific Chemicals Supplier No. 451242500

Chemical Identifiers

CAS 162696-31-3
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.28
InChI Key CSPJUZSNJFANNU-UHFFFAOYSA-N
Synonym 3-boc-aminoethylazetidine, 3-n-boc-aminoethyl azetidine, tert-butyl 2-azetidin-3-yl ethylcarbamate, tert-butyl 2-azetidin-3-yl ethyl carbamate, tert-butyl n-2-azetidin-3-yl ethyl carbamate, 3-2-n-boc-aminoethyl azetidine, tert-butyl 2-3-azetidinyl ethylcarbamate, 2-azetin-3-yl-1-n-boc ethylamine, tert-butyl 2-azetidin-3-yl ethylcarbamate hydrochloride, carbamic acid, n-2-3-azetidinyl ethyl-, 1,1-dimethylethyl ester
PubChem CID 37818431
IUPAC Name tert-butyl N-[2-(azetidin-3-yl)ethyl]carbamate
SMILES CC(C)(C)OC(=O)NCCC1CNC1

Specifications

Melting Point 84°C to 86°C
Assay Percent Range 95% min. (HPLC)
Quantity 250 mg
Formula Weight 200.28
Percent Purity 96%
Chemical Name or Material 3-(N-BOC)-(2-aminoethyl)azetidine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.