Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-n-Octylthiophene 98.0+%, TCI America™
SDP

Catalog No. O021325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O021325G 25 g
O02131G 1 g
O02135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. O021325G Supplier TCI America Supplier No. O021325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 65016-62-8
Molecular Formula C12H20S
Molecular Weight (g/mol) 196.352
MDL Number MFCD00085281
InChI Key WQYWXQCOYRZFAV-UHFFFAOYSA-N
Synonym 3-n-octylthiophene, thiophene, 3-octyl, acmc-1bfcd, 3-octylthiophene, ksc491i4b, bidd:gt0691
PubChem CID 566852
IUPAC Name 3-octylthiophene
SMILES CCCCCCCCC1=CSC=C1

Specifications

Color Colorless
Boiling Point 106°C
Quantity 25 g
Formula Weight 196.35
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-n-Octylthiophene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.