Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-[N-Tris(hydroxymethyl)methylamino]-2-hydroxypropanesulfonic Acid 99.0+%, TCI America™
SDP

Catalog No. T136425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T136425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T136425G Supplier TCI America Supplier No. T136425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

[Good's buffer component for biological research]

Chemical Identifiers

CAS 68399-81-5
Molecular Formula C7H17NO7S
Molecular Weight (g/mol) 259.273
MDL Number MFCD00038352
InChI Key RZQXOGQSPBYUKH-UHFFFAOYSA-N
Synonym tapso, 3-1,3-dihydroxy-2-hydroxymethyl propan-2-yl amino-2-hydroxypropane-1-sulfonic acid, n-tris hydroxymethyl methyl-3-amino-2-hydroxypropanesulfonic acid, 2-hydroxy-3-tris hydroxymethyl methylamino-1-propanesulfonic acid, 3-n-tris hydroxymethyl methylamino-2-hydroxypropane-sulfonic acid, tapso sodium salt, 1-propanesulfonic acid, 2-hydroxy-3-2-hydroxy-1,1-bis hydroxymethyl ethyl amino
PubChem CID 109334
IUPAC Name 3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]-2-hydroxypropane-1-sulfonic acid
SMILES C(C(CS(=O)(=O)O)O)NC(CO)(CO)CO

Specifications

Color White
Quantity 25 g
Formula Weight 259.27
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-[N-Tris(hydroxymethyl)methylamino]-2-hydroxypropanesulfonic Acid [Good′s buffer component fo
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.