Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Nitro-L-tyrosine 98.0+%, TCI America™
SDP

Catalog No. N090525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N090525G 25 g
N09055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N090525G Supplier TCI America Supplier No. N090525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 621-44-3
Molecular Formula C9H10N2O5
Molecular Weight (g/mol) 226.19
MDL Number MFCD00007123
InChI Key FBTSQILOGYXGMD-MDOHGIEYNA-N
Synonym 3-nitro-l-tyrosine, 3-nitrotyrosine, h-tyr 3-no2-oh, l-3-nitrotyrosine, tyrosine, 3-nitro, l-tyrosine, 3-nitro, meta-nitro-tyrosine, unii-7coy1ha6hk, tyrosine, 3-nitro-, l, 3-mononitrotyrosine
PubChem CID 65124
ChEBI CHEBI:44454
IUPAC Name (2S)-2-amino-3-(4-hydroxy-3-nitrophenyl)propanoic acid
SMILES N[C@@H](CC1=CC=C(O)C(=C1)[N+]([O-])=O)C(O)=O

Specifications

Melting Point 224°C
Color Yellow
Quantity 25 g
Formula Weight 226.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Nitro-L-tyrosine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.