Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Nitro-o-phenylenediamine, 98%

Catalog No. AAB2020106
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2020106 5 g
AAB2020103 1 g
AAB2020114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2020106 Supplier Thermo Scientific Chemicals Supplier No. B2020106
Add to Cart

CAS: 3694-52-8 | C6H7N3O2 | 153.141 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3694-52-8
Molecular Formula C6H7N3O2
Molecular Weight (g/mol) 153.141
MDL Number MFCD00007722
InChI Key IOCXBXZBNOYTLQ-UHFFFAOYSA-N
Synonym 3-nitro-1,2-phenylenediamine, 3-nitro-o-phenylenediamine, 1,2-diamino-3-nitrobenzene, 3-nitrophenylenediamine, 2-amino-6-nitroaniline, 1,2-benzenediamine, 3-nitro, 3-nitro-1,2-benzenediamine, o-phenylenediamine, 3-nitro, unii-wrz63l50zk, ccris 5427
PubChem CID 4359525
IUPAC Name 3-nitrobenzene-1,2-diamine
SMILES C1=CC(=C(C(=C1)[N+](=O)[O-])N)N

Specifications

Melting Point 157°C to 162°C
Quantity 5 g
Beilstein 909344
Formula Weight 153.14
Percent Purity 98%
Chemical Name or Material 3-Nitro-o-phenylenediamine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 223-013-5
RTECSNumber ST2974000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.