Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Nitro-o-xylene, 99%

Click to view available options
Quantity:
100g
500g
83-41-0
C8H9NO2
151.165
MFCD00007162
FVHAWXWFPBPFOS-UHFFFAOYSA-N
3-nitro-o-xylene,2,3-dimethylnitrobenzene,nitroxylene,o-xylene, 3-nitro,benzene, 1,2-dimethyl-3-nitro,xylene, ar-nitro,benzene, dimethylnitro,unii-49gnx4cy5w,ccris 3117,49gnx4cy5w
6739
1,2-dimethyl-3-nitrobenzene
CC1=C(C(=CC=C1)[N+](=O)[O-])C
83-41-0
C8H9NO2
151.165
MFCD00007162
FVHAWXWFPBPFOS-UHFFFAOYSA-N
3-nitro-o-xylene,2,3-dimethylnitrobenzene,nitroxylene,o-xylene, 3-nitro,benzene, 1,2-dimethyl-3-nitro,xylene, ar-nitro,benzene, dimethylnitro,unii-49gnx4cy5w,ccris 3117,49gnx4cy5w
6739
1,2-dimethyl-3-nitrobenzene
CC1=C(C(=CC=C1)[N+](=O)[O-])C
Flash Point 107°C (224°F)
Quantity 500g
Melting Point 13°C to 15°C
Boiling Point 244°C to 246°C
UN Number UN1665
Beilstein 2045105
Formula Weight 151.17
Percent Purity 99%
Chemical Name or Material 3-Nitro-o-xylene

GHS H Statement
H311-H331
Toxic in contact with skin.
Toxic if inhaled.

GHS P Statement
P261-P280-P361-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITROXYLENES, LIQUID

TSCA : TSCA