Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Nitro-o-xylene, 99%

Catalog No. p-7027864
Click to view available options
Quantity:
100 g
500 g

CAS: 83-41-0 | C8H9NO2 | 151.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83-41-0
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00007162
InChI Key FVHAWXWFPBPFOS-UHFFFAOYSA-N
Synonym 3-nitro-o-xylene, 2,3-dimethylnitrobenzene, nitroxylene, o-xylene, 3-nitro, benzene, 1,2-dimethyl-3-nitro, xylene, ar-nitro, benzene, dimethylnitro, unii-49gnx4cy5w, ccris 3117, 49gnx4cy5w
PubChem CID 6739
IUPAC Name 1,2-dimethyl-3-nitrobenzene
SMILES CC1=CC=CC(=C1C)[N+]([O-])=O

Specifications

Melting Point 13°C to 15°C
Density 1.129
Boiling Point 244°C to 246°C
Quantity 100 g
Refractive Index 1.544
UN Number UN1665
Beilstein 2045105
Formula Weight 151.17
Percent Purity 99%
Chemical Name or Material 3-Nitro-o-xylene
Hazard Category H312+H332
Hazard Statement GHS H Statement
H311-H331
Toxic in contact with skin.
Toxic if inhaled.
Precautionary Statement P261-P271-P280-P302+P352-P304+P340-P312-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITROXYLENES, LIQUID
EINECSNumber 201-474-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.