Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3'-Nitroacetophenone, 98+%

Catalog No. p-7081930
Click to view available options
Quantity:
250 g
500 g
1000 g

CAS: 121-89-1 | C8H7NO3 | 165.148 g/mol

3'-Nitroacetophenone is used as an intermediate in pharmaceutical industry for making drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3′-Nitroacetophenone is used as an intermediate in pharmaceutical industry for making drugs.

Solubility
Soluble in water 0.75 g/L @ 20°C.

Notes
Store in cool, dry place, in well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-89-1
Molecular Formula C8H7NO3
Molecular Weight (g/mol) 165.148
MDL Number MFCD00007259
InChI Key ARKIFHPFTHVKDT-UHFFFAOYSA-N
Synonym 3'-nitroacetophenone, 3-nitroacetophenone, m-nitroacetophenone, 1-3-nitrophenyl ethanone, ethanone, 1-3-nitrophenyl, acetophenone, 3'-nitro, m-acetylnitrobenzene, usaf ma-1, methyl 3-nitrophenyl ketone, 3-nitroacetofenon
PubChem CID 8494
IUPAC Name 1-(3-nitrophenyl)ethanone
SMILES CC(=O)C1=CC(=CC=C1)[N+](=O)[O-]

Specifications

Melting Point 76°C to 80°C
Boiling Point 202°C
Odor Slightly sweetish
Quantity 250 g
Beilstein 743002
Solubility Information Soluble in water 0.75g/L @ 20°C.
Formula Weight 165.15
Percent Purity ≥98%
Chemical Name or Material 3'-Nitroacetophenone
EINECSNumber 204-504-3
RTECSNumber AM9625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.