Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Nitrobenzoic acid, 99%

Catalog No. AC128455000
Click to view available options
Quantity:
100 g
500 g
5 kg

CAS: 121-92-6 | C7H5NO4 | 167.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-92-6
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00007251
InChI Key AFPHTEQTJZKQAQ-UHFFFAOYSA-N
Synonym 3-nitrobenzoic acid, m-nitrobenzoic acid, benzoic acid, 3-nitro, benzoic acid, m-nitro, meta-nitrobenzoic acid, metanitrobenzoic acid, m-nitrobenzenecarboxylic acid, 3-nitro-benzoic acid, ccris 2335, nitrobenzoic 3-acid
PubChem CID 8497
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 139.0°C to 143.0°C
Color Green to Yellow
Flash Point 190°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula O2NC6H4CO2H
Quantity 500 g
Beilstein 09,376
Fieser 13,231
Merck Index 14,6589
Solubility Information Solubility in water: 3g/L (25°C). Other solubilities: 1g/3mL alcohol - 1g/2mL methanol, 1g/4mL ether - 1g/2.5mL acetone, 1g/18mL chloroform, slightly soluble in benzene and carbon disulfide
Formula Weight 167.12
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material 3-Nitrobenzoic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
EINECSNumber 204-508-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.