Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Nitrobenzonitrile, 98%

Catalog No. AAA1582114
Change view
Click to view available options
Quantity:
10 g
25 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1582114 25 g
AAA1582109 10 g
AAA1582130 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1582114 Supplier Thermo Scientific Chemicals Supplier No. A1582114
Add to Cart

CAS: 619-24-9 | C7H4N2O2 | 148.121 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-24-9
Molecular Formula C7H4N2O2
Molecular Weight (g/mol) 148.121
MDL Number MFCD00007194
InChI Key RUSAWEHOGCWOPG-UHFFFAOYSA-N
Synonym benzonitrile, 3-nitro, m-nitrobenzonitrile, 3-cyanonitrobenzene, m-cyanonitrobenzene, benzonitrile, m-nitro, 3-cyano-1-nitrobenzene, 3-nitro-benzonitrile, ccris 2327, 3-nitrobenzenecarbonitrile, 3-nitrobenzonitril
PubChem CID 12079
IUPAC Name 3-nitrobenzonitrile
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])C#N

Specifications

Melting Point 114°C to 117°C
Boiling Point 165°C (21 mmHg)
Odor Almond-like
Quantity 25 g
UN Number UN3439
Beilstein 637674
Formula Weight 148.12
Percent Purity 98%
Chemical Name or Material 3-Nitrobenzonitrile
Hazard Category H301-H312
Hazard Statement GHS H Statement
H301-H312
Toxic if swallowed.
Harmful in contact with skin.
Precautionary Statement P264b-P270-P280-P301+P310-P302+P352-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 210-587-7
RTECSNumber DI4900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.