Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Nitrobenzyl chloride, 98%

Click to view available options
Quantity:
10g
50g
250g
619-23-8
C7H6ClNO2
171.58
MFCD00007272
APGGSERFJKEWFG-UHFFFAOYSA-N
3-nitrobenzyl chloride,1-chloromethyl-3-nitrobenzene,m-nitrobenzyl chloride,alpha-chloro-3-nitrotoluene,benzene, 1-chloromethyl-3-nitro,toluene, alpha-chloro-m-nitro,.alpha.-chloro-m-nitrotoluene,unii-ojl133898o,3-nitrobenzylchloride,ccris 2324
12078
1-(chloromethyl)-3-nitrobenzene
C1=CC(=CC(=C1)[N+](=O)[O-])CCl
619-23-8
C7H6ClNO2
171.58
MFCD00007272
APGGSERFJKEWFG-UHFFFAOYSA-N
3-nitrobenzyl chloride,1-chloromethyl-3-nitrobenzene,m-nitrobenzyl chloride,alpha-chloro-3-nitrotoluene,benzene, 1-chloromethyl-3-nitro,toluene, alpha-chloro-m-nitro,.alpha.-chloro-m-nitrotoluene,unii-ojl133898o,3-nitrobenzylchloride,ccris 2324
12078
1-(chloromethyl)-3-nitrobenzene
C1=CC(=CC(=C1)[N+](=O)[O-])CCl
Quantity 250g
Melting Point 43°C to 47°C
Flash Point >110°C (230°F)
Boiling Point 150°C (3mmHg)
UN Number UN3261
Beilstein 742794
Formula Weight 171.58
Percent Purity 98%
Chemical Name or Material 3-Nitrobenzyl chloride

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 210-586-1

RTECSNumber : XS9090000

TSCA : Yes

Recommended Storage : Ambient temperatures