Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3-Nitrobenzyl Chloride 99.0+%, TCI America™

Click to view available options
Quantity:
5 g
25 g
Specifications
Specifications
Melting Point | 46°C |
Color | Yellow |
Boiling Point | 174°C |
Quantity | 25 g |
UN Number | 3261 |
Formula Weight | 171.58 |
Percent Purity | ≥99.0% (GC) |
Physical Form | Crystalline Powder |
Chemical Name or Material | 3-Nitrobenzyl Chloride |
Safety and Handling
EINECSNumber | (3)-0449 |
RTECSNumber | XS9090000 |
TSCA | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction