Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Nitrocinnamic Acid 98.0+%, TCI America™
SDP

Catalog No. N035410G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N035410G 10 g
N03545G 5 g
N035425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N035410G Supplier TCI America Supplier No. N035410G

Chemical Identifiers

CAS 555-68-0
Molecular Formula C9H7NO4
Molecular Weight (g/mol) 193.158
MDL Number MFCD00007277
InChI Key WWXMVRYHLZMQIG-SNAWJCMRSA-N
Synonym 3-nitrocinnamic acid, m-nitrocinnamic acid, 3-nitro-cinnamic acid, cinnamic acid, m-nitro, trans-3-nitrocinnamic acid, 3-nitrobenzenepropenoic acid, e-3-3-nitrophenyl-2-propenoic acid, 2e-3-3-nitrophenyl prop-2-enoic acid, 2e-3-3-nitrophenyl acrylic acid, unii-z5wu65j930
PubChem CID 687126
ChEBI CHEBI:60706
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])C=CC(=O)O

Specifications

Melting Point 205°C
Color White-Yellow
Quantity 10 g
Formula Weight 193.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Nitrocinnamic Acid
RTECSNumber GE0352000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.