Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Nitrophthalonitrile 98.0+%, TCI America™
SDP

Catalog No. N056625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N056625G 25 g
N05665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N056625G Supplier TCI America Supplier No. N056625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 51762-67-5
Molecular Formula C8H3N3O2
Molecular Weight (g/mol) 173.13
MDL Number MFCD00191558
InChI Key UZJZIZFCQFZDHP-UHFFFAOYSA-N
Synonym 3-nitrophthalonitrile, 2,3-dicyanonitrobenzene, 1,2-benzenedicarbonitrile, 3-nitro, 3-nitrophthalodinitrile, 1,2-dicyano-3-nitrobenzene, 3-nitro-1,2-benzenedicarbonitrile, nitrophthalonitrile, 3-nitro-phthalonitrile, 3-nitrophthalonitrile?, acmc-209kvo
PubChem CID 162652
IUPAC Name 3-nitrobenzene-1,2-dicarbonitrile
SMILES [O-][N+](=O)C1=CC=CC(C#N)=C1C#N

Specifications

Melting Point 163°C
Quantity 25 g
Formula Weight 173.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Nitrophthalonitrile
RTECSNumber CZ1953200
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.