Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxalyl chloride, 98%

Catalog No. AC129610010
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 79-37-8 | C2Cl2O2 | 126.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent, Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-37-8, 75-44-5, 76-02-8
Molecular Formula C2Cl2O2
Molecular Weight (g/mol) 126.93
MDL Number MFCD00000704
InChI Key CTSLXHKWHWQRSH-UHFFFAOYSA-N
Synonym oxalyl chloride, ethanedioyl dichloride, oxalic dichloride, oxaloyl chloride, oxalic acid dichloride, oxalic acid chloride, ethanedioyl chloride, oxalylchloride, unii-r4y96317dw, cocl 2
PubChem CID 65578
IUPAC Name oxalyl dichloride
SMILES C(=O)(C(=O)Cl)Cl

Specifications

Melting Point -12.0°C
Color Colorless to Yellow
Density 1.4780g/mL
Boiling Point 63.0°C to 64.0°C (763.0 mmHg)
Flash Point >100°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4285 to 1.4305
Linear Formula ClCOCOCl
Quantity 1 kg
Beilstein 02, 542
Fieser 01,767; 02,301; 03,216; 04,361; 05,481; 06,424; 07,257; 08,366; 11,379; 14,150; 16,149; 17,241
Specific Gravity 1.478
Merck Index 15, 7013
Solubility Information Solubility in water: reacts. Other solubilities: soluble in the main common organic solvents :, methylene chloride; dichloroethane; chlorobenzene;, ethylacetate; hexane; acetone; toluene; ether;, benzene; chloroform and ether
Formula Weight 126.93
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Oxalyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if inhaled.
Toxic if swallowed.
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts violently with water.
Contact with w
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Handle under inert gas.
Protect from moisture.
Brush off loose particles from skin.
Immerse in cool water/wrap in wet bandages.
IF INHA
EINECSNumber 201-200-2
RTECSNumber KI2950000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.