Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3-OHKYN Antibody (P3UI), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB100597F
Description
3-OHKYN Monoclonal specifically detects 3-OHKYN in Human, All Species samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
3-OHKYN | |
Monoclonal | |
FITC | |
3039Hydroxykynurenine, 3-hydroxykynuenine modification | |
3-OHKYN modified KLH | |
Protein G purified | |
RUO | |
Primary | |
Human, All species | |
Purified |
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
P3UI | |
ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
Mouse | |
66 kDa | |
0.1 mL | |
Neuroscience, Sensory Systems, Vision | |
This is specific for 3-hydroxykynureinine (3OH-Kyn) modifications on proteins. It is likely to react with 3OH-Kyn modification on lysine, histidine and cysteine residues on proteins. It does not react with enzymes involved in the degradation of tryptopha | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction