Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Phenoxybenzyl alcohol, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
13826-35-2
C13H12O2
200.24
MFCD00004636
KGANAERDZBAECK-UHFFFAOYSA-N
3-phenoxybenzyl alcohol, 3-phenoxyphenyl methanol, 3-phenoxybenzylalcohol, m-phenoxybenzyl alcohol, 3-phenoxybenzenemethanol, benzenemethanol, 3-phenoxy, 3-hydroxymethyl diphenyl ether, 3-phenoxybenzylic alcohol, benzyl alcohol, m-phenoxy, 3-pboh
26295
CHEBI:62527
(3-phenoxyphenyl)methanol
C1=CC=C(C=C1)OC2=CC=CC(=C2)CO
13826-35-2
C13H12O2
200.24
MFCD00004636
KGANAERDZBAECK-UHFFFAOYSA-N
3-phenoxybenzyl alcohol, 3-phenoxyphenyl methanol, 3-phenoxybenzylalcohol, m-phenoxybenzyl alcohol, 3-phenoxybenzenemethanol, benzenemethanol, 3-phenoxy, 3-hydroxymethyl diphenyl ether, 3-phenoxybenzylic alcohol, benzyl alcohol, m-phenoxy, 3-pboh
26295
CHEBI:62527
(3-phenoxyphenyl)methanol
C1=CC=C(C=C1)OC2=CC=CC(=C2)CO
Infrared Spectrum Authentic
Specific Gravity 1.1456
Packaging Glass bottle
Quantity 5g
Flash Point 309°C
Density 1.1456g/mL
Refractive Index 1.5920 to 1.5940
Color Colorless or White
Boiling Point 135.0°C to 140.0°C (1.0mmHg)
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5 % min. (GC)
Linear Formula C6H5OC6H4CH2OH
Solubility Information Solubility in water: insoluble in water.
Formula Weight 200.24
Physical Form Solid or Liquid
Percent Purity 98%
Chemical Name or Material 3-Phenoxybenzyl alcohol, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 237-525-1

RTECSNumber : DP0775400

TSCA : TSCA