Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Phenoxypropionic acid

Catalog No. P160015G Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
10 g

Catalog No. P160015G

Supplier: Sigma Aldrich P160015G

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-ethylfuran undergoes tetraphenylporphin-photosensitized oxygenation in non-polar aprotic solvents via (4+2)-cycloaddition of singlet oxygen to yield the corresponding monomeric unsaturated secondary ozonide.2-ethylfuran was used to investigate the gas phase products formed from the cl atoms initiated reactions of 2-ethylfuran by in situ long-path ftir absorption spectroscopy
  • It was also used in the synthesis of 4-oxo-(e)-2-hexenal via ring opening reaction using aqueous n-bromosuccinimide

Specifications

CAS 7170-38-9
Melting Point 98°C to 100°C (lit.)
Boiling Point 235°C to 245°C (lit.)
Molecular Formula C9H10O3
Linear Formula C6H5OCH2CH2CO2H
MDL Number MFCD00002769
Quantity 10 g
Molecular Weight (g/mol) 166.17
Percent Purity 99%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.