Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Phenyl-1-propylamine, 98%

Catalog No. ac130880250
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle

CAS: 2038-57-5 | C9H13N | 135.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2038-57-5
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00008224
InChI Key LYUQWQRTDLVQGA-UHFFFAOYSA-N
Synonym 3-phenylpropylamine, benzenepropanamine, 1-amino-3-phenylpropane, hydrocinnamylamine, gamma-phenylpropylamine, 3-phenyl-n-propylamine, 3-phenyl-1-propylamine, 3-phenyl-1-propanamine, propylamine, 3-phenyl, 3-phenylpropanamine
PubChem CID 16259
IUPAC Name 3-phenylpropan-1-amine
SMILES NCCCC1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to White
Density 0.9500g/mL
Boiling Point 221°C
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 97.5 % min. (GC)
Packaging Glass bottle
Refractive Index 1.5240 to 1.526
Linear Formula C6H5(CH2)3NH2
Quantity 25 g
Beilstein 12, 1145
Specific Gravity 0.95
Formula Weight 135.21
Percent Purity 98%
Physical Form Crystals or Flakes
Chemical Name or Material 3-Phenyl-1-propylamine, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 218-012-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.