Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Phenyl-1-propyne, 97%, stab.

Catalog No. AAH5350014
Click to view available options
Quantity:
5 g
25 g

CAS: 10147-11-2 | C9H8 | 116.163 g/mol

3-Phenyl-1-propyne is used as a starting material in the synthesis of 4-phenyl-2-butyn-1-ol. It is also used in the preparation of beta-hydroxytriazoles and indoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Phenyl-1-propyne is used as a starting material in the synthesis of 4-phenyl-2-butyn-1-ol. It is also used in the preparation of beta-hydroxytriazoles and indoles.

Notes
Incompatible with acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10147-11-2
Molecular Formula C9H8
Molecular Weight (g/mol) 116.163
MDL Number MFCD00134431
InChI Key NGKSKVYWPINGLI-UHFFFAOYSA-N
Synonym 3-phenyl-1-propyne, 2-propynyl benzene, benzylacetylene, prop-2-yn-1-ylbenzene, benzene, 2-propynyl, propargylbenzol, propargylbenzene, benzyl acetylene, 3-phenyl-propyne, 3-phenyl-1-propyn
PubChem CID 575753
IUPAC Name prop-2-ynylbenzene
SMILES C#CCC1=CC=CC=C1

Specifications

Density 0.934
Boiling Point 75°C (20 mmHg)
Flash Point 52°C (126°F)
Refractive Index 1.526
Linear Formula C6H5CH2C≡CH
Quantity 25 g
UN Number UN3295
Formula Weight 116.16
Percent Purity 97%
Chemical Name or Material 3-Phenyl-1-propyne, stabilized
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.