Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Phenyl-2-piperazinone, 97%, Thermo Scientific™

Catalog No. AAH6309906
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6309906 5 g
AAH63099MD 250 mg
AAH6309903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6309906 Supplier Thermo Scientific Chemicals Supplier No. H6309906

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5368-28-5
Molecular Formula C10H12N2O
Molecular Weight (g/mol) 176.22
MDL Number MFCD01871361
InChI Key WKFFHKBGGZHQAX-UHFFFAOYNA-N
Synonym 3-phenyl-2-piperazinone, 3-phenyl-piperazin-2-one, 2-oxo-3-phenylpiperazine, 3-phenyl-2-piperazinene, 3-phenyl-2-oxopiperazine, 3-phenylpiperazine-2-one, 2-piperazinone,3-phenyl, 2-oxo-3-phenyl-piperazine, 3-phenyl-piperazine-2-one, 2-phenyl-3-keto piperazine
PubChem CID 250681
IUPAC Name 3-phenylpiperazin-2-one
SMILES O=C1NCCNC1C1=CC=CC=C1

Specifications

Quantity 5 g
Formula Weight 176.22
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3-Phenyl-2-piperazinone
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.