Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Phenyl-5-isoxazolone +99%

Catalog No. AC130700050
Click to view available options
Quantity:
5 g
Packaging:
Glass Bottle

CAS: 1076-59-1 | C9H7NO2 | 161.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1076-59-1
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00003158
InChI Key IHKNLPPRTQQACK-UHFFFAOYSA-N
Synonym 3-phenyl-5 4h-isoxazolone, 5 4h-isoxazolone, 3-phenyl, 3-phenyl-2-isoxazolin-5-one, 3-phenylisoxazol-5 4h-one, 3-phenyl-4,5-dihydro-1,2-oxazol-5-one, 2-isoxazolin-5-one, 3-phenyl, 3-phenyl-.delta.2-5-isoxazolinone, 3-phenyl-4-hydroisoxazol-5-one, 3-phenyl-1,2-oxazol-5 4h-one, maybridge1_005968
PubChem CID 70641
IUPAC Name 3-phenyl-4H-1,2-oxazol-5-one
SMILES C1C(=NOC1=O)C2=CC=CC=C2

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 150°C to 154°C
Color Beige to Pink
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass Bottle
Quantity 5 g
Beilstein 27, 200
Formula Weight 161.16
Percent Purity ≥99.0%
Physical Form Crystalline Solid
Chemical Name or Material 3-Phenyl-5-isoxazolone, 99+%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.