Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Phenylpropionaldehyde, 95%

Catalog No. aaa1036718
Click to view available options
Quantity:
50 g
250 g
1000 g

CAS: 104-53-0 | C9H10O | 134.178 g/mol

3-Phenylpropionaldehyde is used as a flavoring agent. It is also used in the preparation of cinnamaldehyde, 2-chloro hydrocinnamaldehyde by alfa chlorination and mixture of homopropargyl alcohols. It serves as a precursor for the synthesis of mixture of syn- and anti-beta-hydroxyallylsilanes by hydroboration of allenylsilane. It is also involved in the catalytic asymmetric cyanosilylation to prepare (2S)-2-hydroxy-4-phenylbutanenitrile. Further, it undergoes Henry reaction to form nitroaldols by reacting with nitromethane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Phenylpropionaldehyde is used as a flavoring agent. It is also used in the preparation of cinnamaldehyde, 2-chloro hydrocinnamaldehyde by alfa chlorination and mixture of homopropargyl alcohols. It serves as a precursor for the synthesis of mixture of syn- and anti-beta-hydroxyallylsilanes by hydroboration of allenylsilane. It is also involved in the catalytic asymmetric cyanosilylation to prepare (2S)-2-hydroxy-4-phenylbutanenitrile. Further, it undergoes Henry reaction to form nitroaldols by reacting with nitromethane.

Solubility
Miscible with chloroform, dichloromethane, ethyl acetate, alcohol and ether. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-53-0
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
MDL Number MFCD00007021
InChI Key YGCZTXZTJXYWCO-UHFFFAOYSA-N
Synonym benzenepropanal, 3-phenylpropionaldehyde, hydrocinnamaldehyde, hydrocinnamic aldehyde, dihydrocinnamaldehyde, benzylacetaldehyde, 3-phenyl-1-propanal, 3-phenylpropylaldehyde, 3-phenylpropyl aldehyde, propanal, phenyl
PubChem CID 7707
IUPAC Name 3-phenylpropanal
SMILES C1=CC=C(C=C1)CCC=O

Specifications

Melting Point °C to 42°C
Density 1.017
Boiling Point 222°C to 223°C
Flash Point 95°C (203°F)
Refractive Index 1.523
Linear Formula C6H5(CH2)2CHO
Quantity 50 g
Beilstein 1071910
Sensitivity Air Sensitive
Solubility Information Miscible with chloroform,dichloromethane,ethyl acetate,alcohol and ether. Immiscible with water.
Formula Weight 134.18
Percent Purity 95%
Chemical Name or Material 3-Phenylpropionaldehyde
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 203-211-8
RTECSNumber MW4890000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.