Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Phenylpropionic acid, 99%

Catalog No. aaa1490822
Click to view available options
Quantity:
100 g
500 g

CAS: 501-52-0 | C9H10O2 | 150.177 g/mol

Hydrocinnamic acid is inhibitor of carboxypeptidase. Hydrocinnamic acid was used in the synthesis of three medium-chain acyl-Coenzyme A?s i.e. 3-phenylpropionyl-CoA from mixed anhydrides of fatty acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hydrocinnamic acid is inhibitor of carboxypeptidase. Hydrocinnamic acid was used in the synthesis of three medium-chain acyl-Coenzyme A′s i.e. 3-phenylpropionyl-CoA from mixed anhydrides of fatty acids.

Solubility
Soluble in water

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 501-52-0
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00002771
InChI Key XMIIGOLPHOKFCH-UHFFFAOYSA-N
Synonym hydrocinnamic acid, 3-phenylpropionic acid, benzylacetic acid, benzenepropanoic acid, dihydrocinnamic acid, benzenepropionic acid, phenylpropanoic acid, 3-phenyl-propionic acid, phenylpropanoate, beta-phenylpropionic acid
PubChem CID 107
ChEBI CHEBI:28631
IUPAC Name 3-phenylpropanoic acid
SMILES C1=CC=C(C=C1)CCC(=O)O

Specifications

Melting Point 47°C to 50°C
Density 1.071
Boiling Point 280°C
Flash Point >110°C (230°F)
Quantity 100 g
Beilstein 907515
Merck Index 14,4784
Solubility Information Soluble in water
Formula Weight 150.18
Percent Purity 99%
Chemical Name or Material 3-Phenylpropionic acid
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 207-924-5
RTECSNumber DA8600000
TSCA Yes

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.