Learn More
3-Phenylpyridine, 95%
Description
3-Phenylpyridine is a useful synthetic intermediate. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3-Phenylpyridine is a useful synthetic intermediate. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in chloroform, dichloromethane and ethyl acetate.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.

Specifications
Specifications
Density | 1.085 |
Boiling Point | 272°C |
Flash Point | >110°C (230°F) |
Refractive Index | 1.616 |
Quantity | 5 g |
Beilstein | 110400 |
Solubility Information | Soluble in chloroform,dichloromethane and ethyl acetate. |
Formula Weight | 155.2 |
Percent Purity | 97% |
Chemical Name or Material | 3-Phenylpyridine |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 213-762-6 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.