Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Propyl gallate, 98%
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 146.0°C to 150.0°C |
Color | White to Beige |
Loss on Drying | 0.5% max. (110°C, 4 hrs) |
Infrared Spectrum | Authentic |
Packaging | Plastic bottle |
Quantity | 2.5 kg |
Merck Index | 15, 7971 |
Solubility Information | Solubility in water: 0.35g/100 mL (25°C). Other solubilities: 103g/100 g alcohol,83g/100 g ether,soluble in aceton,propyleenglycol and,polysorbaat 8 |
Formula Weight | 212.2 |
Percent Purity | 98% |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Warning |
Hazard Statement | GHS H Statement: May cause an allergic skin reaction. Harmful if swallowed. |
Precautionary Statement | GHS P Statement: Wear eye protection/face protection. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
EINECSNumber | 204-498-2 |
RTECSNumber | LW8400000 |
TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction