Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Picolyl chloride hydrochloride, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
500g
Packaging:
Glass bottle
6959-48-4
C6H6ClN·HCl
164.04
MFCD00012818
UZGLOGCJCWBBIV-UHFFFAOYSA-N
3-chloromethyl pyridine hydrochloride,3-chloromethylpyridine hydrochloride,3-picolyl chloride hydrochloride,3-chloromethyl pyridine hcl,ccris 140,pyridine, 3-chloromethyl-, hydrochloride,3-pyridylmethyl chloride hydrochloride,unii-bqa8fyp4yn,3-chloromethyl pyridinium chloride,3-chloromethyl pyridine.hcl
23394
3-(chloromethyl)pyridine;hydrochloride
C1=CC(=CN=C1)CCl.Cl
6959-48-4
C6H6ClN·HCl
164.04
MFCD00012818
UZGLOGCJCWBBIV-UHFFFAOYSA-N
3-chloromethyl pyridine hydrochloride,3-chloromethylpyridine hydrochloride,3-picolyl chloride hydrochloride,3-chloromethyl pyridine hcl,ccris 140,pyridine, 3-chloromethyl-, hydrochloride,3-pyridylmethyl chloride hydrochloride,unii-bqa8fyp4yn,3-chloromethyl pyridinium chloride,3-chloromethyl pyridine.hcl
23394
3-(chloromethyl)pyridine;hydrochloride
C1=CC(=CN=C1)CCl.Cl
Melting Point 137°C to 143°C
Quantity 5g
Color White to Beige
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Solubility Information Solubility in water: 820g/L
Formula Weight 164.04
Physical Form Crystalline Needles or Powder
Percent Purity 99%
Chemical Name or Material 3-Picolyl chloride hydrochloride
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 230-150-4