Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-pyr-Cytisine, Tocris Bioscience™

Catalog No. p-4816926
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

High affinity, partial agonist of α4β2 receptors

High affinity α4β2 partial agonist (Ki values are 0.91, 119 and 1100 nM for α4β2, α3β4 and α7 receptors respectively). Shows little activity at α3β4 and α7 receptors. Exhibits antidepressant-like effects in mouse models of antidepressant efficacy.

Chemical Identifiers

CAS 948027-43-8
Molecular Formula C16H17N3O
Molecular Weight (g/mol) 267.332
InChI Key GOYASQWDLRLAPK-WCQYABFASA-N
Synonym 3-pyr-cytisine, 1r,5s-1,2,3,4,5,6-hexahydro-9-3-pyridinyl-1,5-methano-8h-pyrido 1,2-a 1,5 diazocin-8-one
PubChem CID 16731778
SMILES C1C2CNCC1C3=CC=C(C(=O)N3C2)C4=CN=CC=C4

Specifications

Quantity 10 mg
Formula Weight 267.33
Percent Purity >98%
Chemical Name or Material 3-pyr-Cytisine
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.