Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Pyridinecarboxaldehyde, 98%

Catalog No. AC131831000
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 500-22-1 | C6H5NO | 107.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 500-22-1
Molecular Formula C6H5NO
Molecular Weight (g/mol) 107.11
InChI Key QJZUKDFHGGYHMC-UHFFFAOYSA-N
Synonym 3-pyridinecarboxaldehyde, nicotinaldehyde, 3-pyridinaldehyde, pyridine-3-carboxaldehyde, 3-pyridylcarboxaldehyde, 3-pyridylaldehyde, nicotinealdehyde, 3-formylpyridine, pyridine-3-aldehyde, nicotinic aldehyde
PubChem CID 10371
ChEBI CHEBI:28345
IUPAC Name pyridine-3-carbaldehyde
SMILES C1=CC(=CN=C1)C=O

Specifications

Color Yellow to Brown
Density 1.1420g/mL
Boiling Point 95.0°C to 97.0°C (15.0 mmHg)
Flash Point 84°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5480 to 1.5500
Linear Formula (C5H4N)COH
Quantity 100 g
Beilstein 21,I,288
Specific Gravity 1.142
Solubility Information Solubility in water: miscible. Other solubilities: soluble in most organic solvents
Formula Weight 107.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Pyridinecarboxaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
Causes severe skin burns and eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER
EINECSNumber 207-900-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.