Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Pyridinemethanol, 98%

Catalog No. AAA103810B
Click to view available options
Quantity:
50 g
250 g
1000 g

CAS: 100-55-0 | C6H7NO | 109.13 g/mol

3-Pyridinemethanol is a useful synthetic intermediate. Among other uses, it has been used in the synthesis of histone deacetylase inhibitors. It is also used to solubilize riboflavin and as a peripheral vasodilator; alcohol analog of nicotinic acid; a direct-acting peripheral vasodilator that is used in vasospasm and threatened gangrene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Fully miscible in water.

Notes
Hygroscopic. Store away from oxidizing agents and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-55-0
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.13
MDL Number MFCD00006407
InChI Key MVQVNTPHUGQQHK-UHFFFAOYSA-N
Synonym 3-pyridinemethanol, nicotinyl alcohol, 3-hydroxymethyl pyridine, 3-pyridylmethanol, nicotinic alcohol, 3-pyridylcarbinol, roniacol, pyridylcarbinol, 3-pyridinylmethanol, pyridine-3-carbinol
PubChem CID 7510
IUPAC Name pyridin-3-ylmethanol
SMILES OCC1=CC=CN=C1

Specifications

Melting Point -8°C to -6°C
Density 1.13
Boiling Point 154°C (28 mmHg)
Flash Point 149°C (300°F)
Refractive Index 1.545
Quantity 1000 g
Beilstein 107851
Sensitivity Hygroscopic
Merck Index 14,6527
Solubility Information Fully miscible in water.
Formula Weight 109.13
Percent Purity 98%
Chemical Name or Material 3-Pyridinemethanol
Show More Show Less
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 202-864-6
RTECSNumber UT4690000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.