Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(3-Pyrimidin-2-ylphenyl)methanol, 97%, Thermo Scientific™

Catalog No. p-3527228
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle

Catalog No. CC56309DA

Supplier: Thermo Scientific Chemicals CC56309DA

Only null left
Add to Cart

Chemical Identifiers

CAS 892502-12-4
Molecular Formula C11H10N2O
Molecular Weight (g/mol) 186.21
MDL Number MFCD09064977
InChI Key WMNXGJMCXOLBBV-UHFFFAOYSA-N
Synonym 3-pyrimidin-2-ylphenyl methanol, 3-pyrimidin-2-yl phenyl methanol, benzenemethanol,3-2-pyrimidinyl, 3-pyrimidin-2-yl-benzyl alcohol, benzenemethanol, 3-2-pyrimidinyl, 3-pyrimidin-2-ylphenyl methan-1-ol
PubChem CID 24229565
SMILES OCC1=CC=CC(=C1)C1=NC=CC=N1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name (3-pyrimidin-2-ylphenyl)methanol
Formula Weight 186.21
Percent Purity 97%
Chemical Name or Material (3-Pyrimidin-2-ylphenyl)methanol
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.