Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Pyrrolidinol, 97%

Catalog No. p-4089948
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

CAS: 40499-83-0 | C4H9NO | 87.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40499-83-0
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00005256
InChI Key JHHZLHWJQPUNKB-UHFFFAOYNA-N
Synonym 3-pyrrolidinol, 3-hydroxypyrrolidine, dl-3-pyrrolidinol, 3-hydroxy pyrrolidine, dl-3-hydroxypyrrolidine, rs-3-hydroxypyrrolidine, 3-hydroxypyrrolidinehydrochloride, pyrrolidin-3ol, pyrrolidin-3-o, pyrrolidine-3-ol
PubChem CID 98210
IUPAC Name pyrrolidin-3-ol
SMILES OC1CCNC1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0760g/mL
Boiling Point 179°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4881
Quantity 1 g
Specific Gravity 1.076
Formula Weight 87.12
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Pyrrolidinol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
EINECSNumber 254-944-5
Recommended Storage Product discolors to brown on storage

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.