Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Quinuclidinol, 98+%, Thermo Scientific Chemicals

Catalog No. AAB2150314
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2150314 25 g
AAB2150306 5 g
AAB2150322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2150314 Supplier Thermo Scientific Chemicals Supplier No. B2150314
Add to Cart

3-Quinuclidinol acts as a chiral building block for various antimuscarinic agents and other active pharmaceutical ingredients. It serves as a catalyst for condensation of methyl vinyl ketone with aldehydes. It is also used as a reagent for cleavage of beta-keto and vinylogous beta-keto esters. Further, it plays an important role as a synthon in the preparation of cholinergic receptor ligands and anesthetics. In addition to this, it is used in chemoselective alfa-iodination of acrylic esters through Morita-Baylis-Hillman reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Quinuclidinol acts as a chiral building block for various antimuscarinic agents and other active pharmaceutical ingredients. It serves as a catalyst for condensation of methyl vinyl ketone with aldehydes. It is also used as a reagent for cleavage of beta-keto and vinylogous beta-keto esters. Further, it plays an important role as a synthon in the preparation of cholinergic receptor ligands and anesthetics. In addition to this, it is used in chemoselective alfa-iodination of acrylic esters through Morita-Baylis-Hillman reaction.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1619-34-7
Molecular Formula C7H13NO
Molecular Weight (g/mol) 127.187
MDL Number MFCD00151326
InChI Key IVLICPVPXWEGCA-UHFFFAOYSA-N
Synonym 3-quinuclidinol, quinuclidin-3-ol, 3-hydroxyquinuclidine, 1-azabicyclo 2.2.2 octan-3-ol, quinuclidinol, 1-aza-bicyclo 2.2.2 octan-3-ol, 3-hydroxy-1-azabicyclo 2.2.2 octane, 3-quinuclidinol dl-form, +--3-quinuclidinol, quinuclidine-3-ol
PubChem CID 15381
ChEBI CHEBI:115239
IUPAC Name 1-azabicyclo[2.2.2]octan-3-ol
SMILES C1CN2CCC1C(C2)O

Specifications

Melting Point 217°C to 224°C
pH 10 to 12
Quantity 25 g
UN Number UN3263
Beilstein 104327
Sensitivity Air Sensitive
Merck Index 14,8082
Solubility Information Soluble in water.
Formula Weight 127.19
Percent Purity ≥98%
Chemical Name or Material 3-Quinuclidinol
Show More Show Less
Hazard Category H314-H500
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 216-578-4
RTECSNumber VD6191700
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.