Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Quinuclidinone hydrochloride, 99%, Thermo Scientific Chemicals

Catalog No. p-2716119
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1193-65-3
Molecular Formula C7H11NO·HCl
Molecular Weight (g/mol) 161.63
MDL Number MFCD00137391
InChI Key RFDPHKHXPMDJJD-UHFFFAOYSA-N
Synonym 3-quinuclidinone hydrochloride, quinuclidin-3-one hydrochloride, 3-quinuclidone hydrochloride, 1-azabicyclo 2.2.2 octan-3-one hydrochloride, 3-quinuclidnone hydrochloride, 1-azabicyclo 2.2.2 octan-3-one, hydrochloride, 1-aza-3-oxobicyclo 2.2.2 octane, quinuclidin-3-one, chloride, 1-azabicyclo 2.2.2 octan-3-one, hydrochloride 1:1
PubChem CID 102019
IUPAC Name 1-azabicyclo[2.2.2]octan-3-one;hydrochloride
SMILES C1CN2CCC1C(=O)C2.Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point >300.0°C
Color White
Loss on Drying 0.5% max. (100°C, 2 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98.5 to 101.5% (On dry substance) (Argentometry)
Packaging Plastic bottle
Quantity 25 g
Solubility Information Solubility in water: soluble
Formula Weight 161.63
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 3-Quinuclidinone hydrochloride, 99%
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 214-776-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.