Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Sulfopropyl Methacrylate Potassium Salt 97.0+%, TCI America™
SDP

Catalog No. M115825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M115825G 25 g
M1158500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M115825G Supplier TCI America Supplier No. M115825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 31098-21-2
Molecular Formula C7H11KO5S
Molecular Weight (g/mol) 246.318
MDL Number MFCD00010139
InChI Key PNOXUQIZPBURMT-UHFFFAOYSA-M
Synonym Potassium 3-(Methacryloyloxy)propanesulfonate, Methacrylic Acid 3-Sulfopropyl Ester Potassium Salt
PubChem CID 23682747
IUPAC Name potassium;3-(2-methylprop-2-enoyloxy)propane-1-sulfonate
SMILES CC(=C)C(=O)OCCCS(=O)(=O)[O-].[K+]

Specifications

Melting Point 295°C
Color White
Quantity 25 g
Formula Weight 246.32
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Sulfopropyl Methacrylate Potassium Salt
EINECSNumber (2)-1613&(2)-2829
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.