Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(tert-Butoxycarbonylamino)-1-propanol 96.0+%, TCI America™
SDP

Catalog No. H090025ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H090025ML 25 mL
H09005ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H090025ML Supplier TCI America Supplier No. H090025ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 58885-58-8
Molecular Formula C8H17NO3
Molecular Weight (g/mol) 175.228
MDL Number MFCD00191883
InChI Key XDJCYKMWJCYQJM-UHFFFAOYSA-N
Synonym 3-boc-amino-1-propanol, tert-butyl n-3-hydroxypropyl carbamate, boc-beta-ala-ol, 3-tert-butoxycarbonylamino-1-propanol, tert-butyl 3-hydroxypropylcarbamate, 3-n-boc-amino propanol, tert-butyl 3-hydroxypropyl carbamate, n-3-hydroxypropyl carbamic acid tert-butyl ester, 3-hydroxy-propyl-carbamic acid tert-butyl ester
PubChem CID 3838134
IUPAC Name tert-butyl N-(3-hydroxypropyl)carbamate
SMILES CC(C)(C)OC(=O)NCCCO

Specifications

Color Colorless
Quantity 25 mL
Formula Weight 175.23
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(tert-Butoxycarbonylamino)-1-propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.