Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(tert-Butyl)-9H-carbazole 95.0+%, TCI America™
SDP

Catalog No. B50531G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B50531G 1 g
B5053200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B50531G Supplier TCI America Supplier No. B50531G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 22401-74-7
Molecular Formula C16H17N
Molecular Weight (g/mol) 223.319
MDL Number MFCD12024281
InChI Key TYXSZNGDCCGIBO-UHFFFAOYSA-N
PubChem CID 12117342
IUPAC Name 3-tert-butyl-9H-carbazole
SMILES CC(C)(C)C1=CC2=C(C=C1)NC3=CC=CC=C32

Specifications

Melting Point 153°C
Color White
Quantity 1 g
Formula Weight 223.32
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-(tert-Butyl)-9H-carbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.